Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wide Mouth Round Bottles with Heavy Duty Closure Available on GSA/VA Contract for Federal Government customers only.

For handling chemicals both in liquid and in powder forms

Manufacturer:  Bel-Art™ F109062000

 View more versions of this product

Catalog No. 03-409-30H


Add to cart

Description

Description

  • For use in storing, shipping and handling materials
  • Available in different volumes ranging from 50mL to 10L

LDPE bottles:

  • Heavy threaded, watertight design screw cap closure with inner seal
  • HDPE bottles:
    • Octagonal shaped
    • More suited for powders

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Specifications

    Specifications

    68 oz., 2000mL
    68 oz.
    50mm
    120mm
    Screw
    2000mL
    Translucent
    LDPE
    255mm
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit