Learn More
Bel-Art™ SP Scienceware™ Write-On™ Colored Label Tapes
Custom manufactured from proprietary materials that resist moisture and most solvents
Manufacturer:  Bel-Art™ 134620005
 View more versions of this product
Catalog No. 11-865-100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Colors correspond with familiar hazard color codes to highlight safety information written on the labels.
  • Adhere to TFE fluoropolymer resin covered materials and any clean surface
  • Can endure a maximum temperature of 247°C (492°F) for 30 minutes
  • Samples stay labeled even after steam autoclaving at 121°C (250°F) or freezing down to -73°C (-99°F)
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Specifications
Specifications
White
0.5 in.
Tape