Learn More
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
Manufacturer:  Bel-Art™ F133530002
 View more versions of this product
Catalog No. 01-815-12B
Add to cart
Description
Description
  • Made of black nylon
  • Hook-and-loop sealing strip on its closing flap
Specifications
Specifications
Security Bag
Used for additional protection from light for X-ray film cassettes that you are currently using
5 × 7 in.
Nylon
Black
X-ray film cassette
12.7 × 17.7cm