Learn More
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags Available on GSA/VA Contract for Federal Government customers only.
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
Manufacturer:  Bel-Art™ F133530000
 View more versions of this product
Catalog No. 01-815-12
Add to cart
Description
Description
  • Made of black nylon
  • Hook-and-loop sealing strip on its closing flap
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Black
X-ray film cassette
38.1 × 45.7cm
Security Bag
Used for additional protection from light for X-ray film cassettes that you are currently using
15 × 18 in.
Nylon