Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art SP Scienceware X-Ray Cassette Security Bags
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
Specifications
Color | Black |
---|---|
Product Type | Security Bag |
For Use With (Application) | Used for additional protection from light for X-ray film cassettes that you are currently using |
For Use With (Equipment) | X-ray film cassette |
Material | Nylon |
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Price | Quantity & Availability | ||||
01-815-12B
|
Bel-Art
F133530002 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Made of black nylon
- Hook-and-loop sealing strip on its closing flap
Specifications
Black | |
Used for additional protection from light for X-ray film cassettes that you are currently using | |
Nylon |
Security Bag | |
X-ray film cassette |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title