Bel-Art™ Micro-Tube Homogenizer: Pestles and Tubes

Chemical and heat resistant, colorant-free

Manufacturer: Bel-Art™ F650009004

 View more versions of this product

Catalog No. 03-421-225

  Check Availability
Add to Cart

    Specifications

    Material Animal and phalate/phalate-ester free polypropylene
    Type Tube
    For Use With (Equipment) Micro-Tube homogenizer system
    Size 1.5mL
    RNase Free Yes
    Dnase Free Yes
    Outer Packaging Bulk pack
    Quantity 100/Pk.

    Matched pestle tips and micro tubes can be used either with homogenizer or with hand processing.

    • Tight tolerances, thorough homogenization and grinding of materials
    • Chemical and heat resistant, colorant-free to eliminate potential leaching of heavy metals into samples
    • Manufactured and packaged in class 10,000 cleanroom
    • Certified DNase, RNase and Pyrogen-free
    • Made of animal and phalate/phalate-ester free polypropylene
    • Available in individual or bulk packs
    Pestles
    • Unique ribbed shaft provides precise finger control of homogenization for shear sensitive samples
    • Fit most motorized mixer adapters
    Tubes
    • Tube bottom 25% thicker bottoms than industry standard
    • Frosted side and cap surfaces for labeling, positive seal snap-cap design and pierceable top

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit