Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Belladonna Extract, Powdered Leaf Extract, USP, 1.15-1.35%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

BE111

Spectrum™ Belladonna Extract, Powdered Leaf Extract, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Quantity 25 kg
CAS Max % 1
Percent Purity 1.15 to 1.35%