Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Belladonna Tincture, USP, 27-33 mg/100 mL, Spectrum™

Click to view available options
Quantity:
20 L
Packaging:
Poly Drum

BE115

Spectrum™ Belladonna Tincture, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Drum
Quantity 20 L
CAS Max % 100%
UN Number UN1170