Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benazepril hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
86541-74-4
C24H29ClN2O5
460.96
MFCD00895734
VPSRQEHTHIMDQM-FKLPMGAJSA-N
benazepril hydrochloride, benazepril hcl, lotensin, cibacen, cibacene, briem, tensanil, zinadril, cibace, labopol
5362123
CHEBI:3012
hydrogen 2-[(3S)-3-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic acid chloride
[H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCC2=CC=CC=C2N(CC(O)=O)C1=O
This item is not returnable. View return policy
86541-74-4
C24H29ClN2O5
460.96
MFCD00895734
VPSRQEHTHIMDQM-FKLPMGAJSA-N
benazepril hydrochloride, benazepril hcl, lotensin, cibacen, cibacene, briem, tensanil, zinadril, cibace, labopol
5362123
CHEBI:3012
hydrogen 2-[(3S)-3-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic acid chloride
[H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCC2=CC=CC=C2N(CC(O)=O)C1=O

Angiotensin-converting enzyme (ACE) inhibitor

Non-peptide angiotensin-converting enzyme (ACE) inhibitor. Reduces blood pressure and myocardial hypertrophy in spontaneous hypertensive rats.
Quantity 50 mg
Formula Weight 460.96
Percent Purity >99%
Chemical Name or Material Benazepril hydrochloride

Recommended Storage : Desiccate at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit