Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitchell Plastics Bench Mat Dispensers

Specifications

Product Type Assay Mat Rack Dispenser
Material Acrylic
Quantity Each
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
05-723-421
View Documents
Mitchell Plastics
MP1610G
Green Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
05-723-422
View Documents
Mitchell Plastics
MP1610R
Red Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Specifications

Specifications

Assay Mat Rack Dispenser
Acrylic
Each
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit