Learn More
Cytiva Benchkote Sheet for ÄKTA Systems PROMO
Protects the top buffer trays from buffer spillages and salt deposits
$7.51 - $17.08
Specifications
Product Type Benchkote Sheets
Material Absorbent Paper, Laminated Polyethylene
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
09-928-149LR1
View Documents
Cytiva
29648076
ÄKTA Avant
Pack of 10 for $17.08
Add to cart
09-928-146LR1
View Documents
Cytiva
29648078
ÄKTA Start
Pack of 10 for $7.51
Add to cart
Description
Description
  • Cover the top buffer tray to protect from buffer spillages and salt deposits
  • Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
  • Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
  • Smooth, absorbent Whatman paper quickly absorbs spills
  • Laminated polyethylene layer prevents flow-through to the working surface
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Convenient and easy to use
Specifications
Specifications
Benchkote Sheets
Absorbent Paper, Laminated Polyethylene