Learn More
Cytiva Benchkote sheet for ÄKTA systems PROMO
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
Specifications
Description For use with ÄKTA go
Material Absorbent paper and laminated polyethylene
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09-920-054LR0
View Documents
Cytiva
29489288
10/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
09-920-056LR0
View Documents
Cytiva
29489289
25/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Why Benchkote sheet for lab ÄKTA systems?
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
  • Smooth, absorbent Whatman™ paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
  • Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
  • The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Specifications
Specifications
For use with ÄKTA go
Absorbent paper and laminated polyethylene