Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Benchkote™ Plus Surface Protector Sheets PROMO

Prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. Whatman™ Benchkote Plus bench liner can also help with the disposal of hazardous materials.

Manufacturer:  GE Healthcare 23016150PM X

Catalog No. 12-007-183APM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Strong: Benchkote Plus bench liner is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote Plus bench paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
Specifications

Specifications

Surface protector
Suitable for saturation with disinfectant
0.75L/m2
482mm
600mm
482 x 580mm (19 x 23 in.)
Polyethylene
19 in.
23 in.
50 Sheets
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit