Learn More
Sartorius™ Benchtop Meters: pHBasic Series Available on GSA/VA Contract for Federal Government customers only.
For basic everyday pH routines
Manufacturer:  Sartorius™ PHBASICKIT
 View more versions of this product
Catalog No. 14-558-064
Add to cart
Description
Description
  • Simple-to-use with classic interface design and intuitive icons
  • Classic no-fuss menus and intuitive operation
  • Works faster due to optimized algorithms with harmonized measuring system of sensor with meter
  • Large LCD displays with simultaneous pH/mV and temperature
  • Reliable with understandable status icons and real time indicators like ElectCheck, and Stability Control
  • Automatic temperature compensation
  • Automatic buffer recognition- up to 16 buffer sets
  • Easy 1-2-3 point Calibration
  • German quality
  • Quick access to electrode calibration data
  • At a glance know if sensor efficiency and calibration integrity are intact
Note: ElectCheck is a sensor evaluation and calibration reminder. The sensor evaluation shows the electrode quality in increments. The integrity criteria are slope, zero point and reaction time. When the time interval has elapsed, an optical and an acoustical signal is activated.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
LCD (Large Displays with Simultaneous pH/mV and Temperature)
3
Sample and Buffer Preparation - R&D environments
pHBasic Kit with PY-P10-2S electrode
2 years
Meter, Power Supply, Cover, Manual, 3 Buffers, Arm, Electrode PY-P10 -2S (3pt Cal and ElectCheck)
Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P10-2S
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021