Learn More
Sartorius™ Benchtop Meters: pHBasic+ Series
For entire pH requirements
Manufacturer:  Sartorius™ PHBASIC(PLUS)BIOKITX
 View more versions of this product
Catalog No. 14-558-070
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Fast and accurate results, easy operation, and capacity to have direct data recall and transfer
  • Full functionality with USB data transmission
  • Recall and output of 5,000 data set memory
  • Easy-to-use menus and intuitive operation sets
  • Sensor efficiency & calibration integrity
  • Clear status icons and indicators
  • Large LCD display
  • Automatic temperature compensation
  • Automatic buffer recognition
  • Programmable data collection
  • 5-point calibration gives a broader range in slope accuracy
  • Features Continuous Monitoring Control CMC, GLP TIME/DATE stamps 
Specifications
Specifications
LCD
5
Sample and Buffer Preparation - Regulated R&D and QC environments - Biological environments:Cell culture, blood separation, microbiology, proteomics genomics, and molecular biology
Data Memory 500 and output
pHBasic+ Bio Kit PY-P28-2S electrode
2 years
Meter, Power Supply, Cover, Arm, 3 Buffer, Electrode: PY-P28-2S (5pt Cal, ElectCheck, USB,CMC)
Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P28-2S
USB
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021