Sartorius™ Benchtop Meters: pHBasic Series
For basic everyday pH routines
Manufacturer: Sartorius™ PHBASICFOODKIT
 View more versions of this product
Catalog No. 14-558-066
  Check Availability
Add to Cart
    Specifications
    Description Meter, Power Supply, Cover, Manual, 3 Buffers, Arm, Electrode: PY-P27-2S(3pt Cal and Electcheck)
    Calibration Points (pH) 3 points
    Display Type Large LCD displays with simultaneous pH/mV and temperature
    Includes Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P27-2S
    For Use With (Application) Sample and Buffer Preparation for non aqueous samples: Media prepartion, Food and Beverage sites, Cosmetics
    Warranty 2 years
    Product Type pHBasic Food Kit with PY-P27-2S electrode
    • Simple-to-use with classic interface design and intuitive icons
    • Classic no-fuss menus and intuitive operation
    • Works faster due to optimized algorithms with harmonized measuring system of sensor with meter
    • Large LCD displays with simultaneous pH/mV and temperature
    • Reliable with understandable status icons and real time indicators like ElectCheck, and Stability Control
    • Automatic temperature compensation
    • Automatic buffer recognition- up to 16 buffer sets
    • Easy 1-2-3 point Calibration
    • German quality
    • Quick access to electrode calibration data
    • At a glance know if sensor efficiency and calibration integrity are intact
    Note: ElectCheck is a sensor evaluation and calibration reminder. The sensor evaluation shows the electrode quality in increments. The integrity criteria are slope, zero point and reaction time. When the time interval has elapsed, an optical and an acoustical signal is activated.
  • Description & Specifications
Related Products
Product Content Feedback