Sartorius™ Benchtop Meters: pHBasic+ Series
For entire pH requirements
Manufacturer: Sartorius™ PHBASIC(PLUS)A
 View more versions of this product
Catalog No. 14-558-067
  Check Availability
Add to Cart
    Specifications
    Description Meter, Power Supply, Cover (5pt Cal, ElectCheck, USB,CMC)
    Calibration Points (pH) 5 points
    Display Type Large LCD display
    Memory Data Memory 500 and output
    Outputs USB Interface
    Includes Power Supply, Manual, 4 AA batteries
    For Use With (Application) Sample and Buffer Preparation - Regulated R&D and QC environments
    Warranty 2 years
    Product Type pHBasic+ Meter only
    • Fast and accurate results, easy operation, and capacity to have direct data recall and transfer
    • Full functionality with USB data transmission
    • Recall and output of 5,000 data set memory
    • Easy-to-use menus and intuitive operation sets
    • Sensor efficiency & calibration integrity
    • Clear status icons and indicators
    • Large LCD display
    • Automatic temperature compensation
    • Automatic buffer recognition
    • Programmable data collection
    • 5-point calibration gives a broader range in slope accuracy
    • Features Continuous Monitoring Control CMC, GLP TIME/DATE stamps 
  • Description & Specifications
Related Products
Product Content Feedback