Learn More
Sartorius Benchtop Meters: pHCore Series
Features stability control, automatic buffer recognition, automatic temperature compensation, AutoCal to ensure accurate measurements
Specifications
For Use With (Application) Sample and Buffer Preparation - Teaching labs and production enivironments
Calibration Points 2
Warranty 2 Years
Display Type LCD
View More Specs
Products 1
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
14-558-061
View Documents
Sartorius
PHCOREKIT
pHCore Kit with Electrode PY-P20-2S Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P20-2S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Easy Key Operation: Mode, Cal, Stability control, OK, Scroll Navigation
  • Large, Easy-To-Read LCD Display instantly shows what samples and system are doing
  • Autocal can be set for easy recognition of daily buffer set
  • German quality
  • Clear Status Icons and Indicators
  • Simultaneous pH and Temp °C or °F
  • Quick Display of Electrode Calibration
  • Recognizes 16 Standard Buffers
  • Seamless 2-point calibration routine
Specifications
Specifications
Sample and Buffer Preparation - Teaching labs and production enivironments
2
2 Years
LCD