Sartorius™ Benchtop Meters: pHCore Series
Features stability control, automatic buffer recognition, automatic temperature compensation, AutoCal to ensure accurate measurements
Manufacturer: Sartorius™ PHCOREA
 View more versions of this product
Catalog No. 14-558-060
  Check Availability
Add to Cart
    Specifications
    Description Meter, Power Supply, Cover, Manual (2pt Cal)
    Calibration Points (pH) 2 points
    Display Type Large LCD display
    Includes Power Supply, Manual, 4 AA batteries
    For Use With (Application) Sample and Buffer Preparation - Teaching labs and production enivironments
    Warranty 2 years
    Product Type pHCore Meter Only
    • Easy Key Operation: Mode, Cal, Stability control, OK, Scroll Navigation
    • Large, Easy-To-Read LCD Display instantly shows what samples and system are doing
    • Autocal can be set for easy recognition of daily buffer set
    • German quality
    • Clear Status Icons and Indicators
    • Simultaneous pH and Temp °C or °F
    • Quick Display of Electrode Calibration
    • Recognizes 16 Standard Buffers
    • Seamless 2-point calibration routine
  • Description & Specifications
Related Products
Product Content Feedback