Learn More
Sartorius™ Benchtop Meters: Pro 15 Series Available on GSA/VA Contract for Federal Government customers only.
For advanced professional pH and ISE analyses
Manufacturer:  Sartorius™ PR15PHKIT
 View more versions of this product
Catalog No. 14-558-073
Add to cart
Description
Description
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Meter, Power Supply, Cover, Arm, 3 Buffers, Manual, Electrode: PY-P11-2S (5pt,GLP)
5
Data Memory 1500 and output
Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments
PR-15 Kit with PY-P11-2S electrode
Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P11-2S
LCD
USB, RS-232
2 years
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021