Learn More
Sartorius Benchtop Meters: Pro 15 Series
For advanced professional pH and ISE analyses
Specifications
For Use With (Application) Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments
Calibration Points 5
Warranty 2 Years
Outputs USB, RS-232
Memory Data Memory 1500 and output
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
14-558-073
View Documents
Sartorius
PR15PHKIT
PR-15 Kit with PY-P11-2S electrode Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P11-2S N/A N/A N/A
14-558-074
View Documents
Sartorius
PR15BIOKIT
PR-15 Bio Kit with PY-P28-2S electrode Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P28-2S N/A N/A N/A
Description
Description
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
Specifications
Specifications
Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments
2 Years
Data Memory 1500 and output
5
USB, RS-232
LCD