Sartorius™ Benchtop Meters: Pro 15 Series
For advanced professional pH and ISE analyses
Manufacturer: Sartorius™ PR15A
 View more versions of this product
Catalog No. 14-558-072
  Check Availability
Add to Cart
    Specifications
    Description Meter, Power supply, Cover, Arm, 3 Buffers (5pt,GLP)
    Calibration Points (pH) 5 points
    Display Type Large LCD display
    Memory Data Memory 1500 and output
    Outputs USB and RS232 Interface
    Includes Power Supply, Manual, 4 AA batteries
    For Use With (Application) Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments
    Warranty 2 years
    Product Type PR-15A Meter Only
    • Exact measurements with wider measuring ranges and better accuracy specifications
    • Advance GLP Documentation with time and date stamps
    • Direct data transmission with RS232 and USB interfaces
    • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
    • Multi-functionality in One
    • Complete Documentation
    • Reliable results with automatic and wireless sensor and user ID recognition
    • User name can be assigned to each measured value
    • Complies with GLP requirements with administrator rights or user identification via electronic ID card
    • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
    • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback