Learn More
Sartorius Benchtop Meters: Pro 20 Series
For advanced professional conductivity analyses
Manufacturer: Sartorius PR20CONKIT
Description
- Exact measurements with wider measuring ranges and better accuracy specifications
- Advance GLP Documentation with time and date stamps
- Direct data transmission with RS232 and USB interfaces
- Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
- Multi-functionality in One
- Complete Documentation
- Reliable results with automatic and wireless sensor and user ID recognition
- User name can be assigned to each measured value
- Complies with GLP requirements with administrator rights or user identification via electronic ID card
- Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
- Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
Specifications
5 | |
Large LCD | |
Sample and Buffer Preparation - Regulated R&D and QC environments: Academic, Wasterwater, Food | |
Data Memory 1500 and output | |
2 Years |
Meter, Power Supply, Cover, Arm, Conductivity Sensor: PY-C12-2S (5pt,GLP) | |
USB, RS232 | |
Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Cell PY-C12-2S | |
PR-20 Conducitivity Kite with PY-C12-2S Electrode |