Learn More
Sartorius Benchtop Meters: Pro 20 Series
For advanced professional conductivity analyses
Specifications
For Use With (Application) Sample and Buffer Preparation - Regulated R&D and QC environments: Academic, Wasterwater, Food
Warranty 2 Years
Outputs USB, RS232
Memory Data Memory 1500 and output
Display Type Large LCD
View More Specs
Products 1
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
14-558-076
View Documents Promotion Details
Sartorius
PR20CONKIT
PR-20 Conducitivity Kite with PY-C12-2S Electrode Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Cell PY-C12-2S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
Specifications
Specifications
Sample and Buffer Preparation - Regulated R&D and QC environments: Academic, Wasterwater, Food
USB, RS232
Large LCD
2 Years
Data Memory 1500 and output