Learn More
Sartorius™ Benchtop Meters: Pro 50 Series
For advanced professional pH, ISE, DO, Conductivity analyses
Manufacturer:  Sartorius™ PR50A
 View more versions of this product
Catalog No. 14-558-077
Add to cart
Description
Description
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
Specifications
Specifications
Meter with ID sensor Card. B&W Display. Menu Structured Handling. Recorder Function. Cond pH, mV, Temp, ISE,DO RS232- USB slave 1BNC . Incl. Power Supply. Cover, Arm,3 Buffers (5pt,GLP)
5 points
Data Memory 1500 and output
Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments
PR-50 Meter Only
Power Supply, Manual, 4 AA batteries
Large LCD display
USB and RS232 Interface
2 years
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021