Learn More
Sartorius™ Benchtop Meters: Pro 50 Series
For advanced professional pH, ISE, DO, Conductivity analyses
$3030.00
Specifications
Calibration Points (pH) 5 points
Warranty 2 years
Outputs USB and RS232 Interface
Memory Data Memory 1500 and output
Display Type Large LCD display
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
14-558-077 View Documents Sartorius™
PR50A
PR-50 Meter Only Power Supply, Manual, 4 AA batteries Each for $3,030.00
Add to cart
14-558-078 View Documents Sartorius™
PR50BIOKIT
PR-50 Bio Kit with PY-P28-2S electrode Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P28-2S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
Specifications
Specifications
5 points
USB and RS232 Interface
Large LCD display
2 years
Data Memory 1500 and output