Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BEND4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | BEND4 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4501
![]() ![]() |
Proteintech Group Inc
247111AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.Specifications
BEND4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P86174, Q6ZU67 | |
389206, 666938 | |
BEND4 Fusion Protein Ag20169 | |
Primary | |
-20°C | |
BEND4 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BEN domain containing 4, BEND4, CCDC4 | |
BEND4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title