Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BEND7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548448

Catalog No. PA548448

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Specifications

Specifications

BEND7
Polyclonal
PBS with 0.09% sodium azide
Q8N7W2, Q8BSV3
BEN domain-containing protein 7, BEND7, C10orf30
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
209645, 222389
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BEND7
Liquid
BEND7
KLH conjugated synthetic peptide between 319-348 amino acids from the C-terminal region of human BEND7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit