Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benidipine hydrochloride, Tocris Bioscience™

Catalog Number 393450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
91599-74-5
C28H32ClN3O6
542.029
KILKDKRQBYMKQX-MIPPOABVSA-N
(4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
656667
5-O-[(3R)-1-benzylpiperidin-3-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OC2CCCN(C2)CC3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl
This item is not returnable. View return policy
91599-74-5
C28H32ClN3O6
542.029
KILKDKRQBYMKQX-MIPPOABVSA-N
(4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
656667
5-O-[(3R)-1-benzylpiperidin-3-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OC2CCCN(C2)CC3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl

Calcium channel blocker

Orally active antihypertensive agent which displays a wide range of activities in vitro and in vivo. Inhibits L-, N- and T-type Ca2+ channels. Also inhibits aldosterone-induced mineralocorticoid receptor activation. Exhibits cardioprotective and antiartherosclerotic effects.
Quantity 50 mg
Formula Weight 542.02
Percent Purity >99%
Chemical Name or Material Benidipine hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit