Learn More
Medchemexpress LLC Benzaldehyde, 2,4,5-trimethoxy- | 4460-86-0 | MFCD00003312 | 99.8% | 196.20 g/mol | C10H12O4 | 500 G

Supplier: Medchemexpress LLC HY100580500G
Asaraldehyde (2,4,5-trimethoxybenzaldehyde) is a naturally occurring aromatic aldehyde used as a research reagent and reported COX-2 inhibitor. It is supplied as a high-purity solid for analytical and biochemical studies and has documented activity against cyclooxygenase-2 at 100 μg/mL.
- Reported COX-2 inhibitor, IC50 = 100 μg/mL.
- Chemical formula C10H12O4.
- Molecular weight 196.20 g/mol.
- High purity (99.8%).
- Provided as a solid suitable for analytical and assay use.
- Available in large pack sizes for bulk laboratory applications.
- Intended for research use only; not for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.