Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzaldehyde dimethyl acetal, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
250mL
1L
2.5L
Packaging:
Glass bottle
1125-88-8
C9H12O2
152.19
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal,dimethoxymethyl benzene,alpha,alpha-dimethoxytoluene,dimethoxyphenylmethane,benzene, dimethoxymethyl,benzaldehydedimethylacetal,dimethoxymethyl-benzene,benzaldehyde, dimethyl acetal,benzaldehyde dimethylacetal,unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC
1125-88-8
C9H12O2
152.19
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal,dimethoxymethyl benzene,alpha,alpha-dimethoxytoluene,dimethoxyphenylmethane,benzene, dimethoxymethyl,benzaldehydedimethylacetal,dimethoxymethyl-benzene,benzaldehyde, dimethyl acetal,benzaldehyde dimethylacetal,unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC
Quantity 100mL
Flash Point 69°C
Specific Gravity 1.01
Color Colorless
Packaging Glass bottle
Refractive Index 1.4920 to 1.4940
Infrared Spectrum Authentic
Density 1.0100g/mL
Boiling Point 87.0°C to 89.0°C (18.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CH(OCH3)2
Beilstein 07,209
Solubility Information Solubility in water: decomposes
Formula Weight 152.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Benzaldehyde dimethyl acetal, 99+%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 214-413-0

RTECSNumber : CU5774000

TSCA : TSCA