Alfa Aesar™ Benzaldoxime, predominantly (E)-isomer, 98%

Manufacturer: Alfa Aesar™ A1205336

Quantity 500g

 View more versions of this product

Catalog No. AAA1205336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 932-90-1
    Molecular Formula C7H7NO
    Formula Weight 121.14
    MDL Number MFCD00002119
    Beilstein 774138
    Synonym Benzaldehyde oxime
    Chemical Name or Material Benzaldoxime, predominantly (E)-isomer

    Specifications

    Formula Weight 121.14
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 932-90-1
    Flash Point 108°C (226°F)
    Quantity 500g
    Beilstein 774138
    Synonym Benzaldehyde oxime
    Melting Point 33° to 35°C
    Boiling Point 118° to 120°C (10mmHg)
    Molecular Formula C7H7NO
    MDL Number MFCD00002119
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 1 Warning
    Chemical Name or Material Benzaldoxime, predominantly (E)-isomer
    Assay 98%

    Description

    Applications
    Oximes are used as a peel-preventing additive in paints and lacquers. It acts as an antioxidant against oxidative drying materials which forms sticky skin with air oxygen. Oximes are used as chemical building block for the synthesis of agrochemicals and pharmaceuticals. In medicine application, Oxime structure is effective in cholinesterase reactivators to treat the poisoning by organophosphates. Oxime is used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

    Solubility
    Slightly soluble in water. Soluble in ethanol and ether.

    Notes
    Store in cool place. Keep away from oxidizing agents. Incompatible with bases, acids, acid anhydrides and acid chlorides.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit