Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzaldoxime, predominantly (E)-isomer, 98%

Manufacturer:  Alfa Aesar™ A1205322

Quantity 100g

 View more versions of this product

Catalog No. AAA1205322


  Check Availability
Add to cart

Specifications

Description

Applications
Oximes are used as a peel-preventing additive in paints and lacquers. It acts as an antioxidant against oxidative drying materials which forms sticky skin with air oxygen. Oximes are used as chemical building block for the synthesis of agrochemicals and pharmaceuticals. In medicine application, Oxime structure is effective in cholinesterase reactivators to treat the poisoning by organophosphates. Oxime is used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

Solubility
Slightly soluble in water. Soluble in ethanol and ether.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases, acids, acid anhydrides and acid chlorides.
Description & Specifications

Chemical Identifiers

932-90-1
121.14
774138
Benzaldoxime, predominantly (E)-isomer
C7H7NO
MFCD00002119
Benzaldehyde oxime

Specifications

932-90-1
121.14
774138
33° to 35°C
100g
Benzaldoxime, predominantly (E)-isomer
C7H7NO
MFCD00002119
Benzaldehyde oxime
118° to 120°C (10mmHg)
108°C (226°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit