Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
5kg
Packaging:
Glass bottle
Plastic Bottle
8001-54-5
C9H13ClN
170.66
MFCD00145757
Alkyl-benzyl-dimethylammonium chloride
8001-54-5
C9H13ClN
170.66
MFCD00145757
Alkyl-benzyl-dimethylammonium chloride
Water 10% Max. (K.F.)
Color Undesignated
Sulfated Ash 0.5% max.
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC16H33
Merck Index 15, 1061
Physical Form Solid or Thick Gel
Percent Purity 95.0 to 104.0% (anhydrous basis)
Chemical Name or Material Benzalkonium chloride
Show More Show Less

GHS P Statement Harmful if swallowed. Very toxic to aquatic life. Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not get in eyes, on skin, or on clothing. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Get medical attention/advice if you feel unwell.

Danger