Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium Chloride, NF, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Drum

B1620, 8001-54-5

Spectrum™ Benzalkonium Chloride, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilizing agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS 8001-54-5
CAS Max % 1
Molecular Formula C9H13ClN
UN Number UN2923
SMILES [Cl-].C[N+](C)(*)CC1=CC=CC=C1
Molecular Weight (g/mol) 170.66
Ignition Residue 0.02
Packaging Drum
Quantity 25 kg