Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium Chloride Solution, NF, 47.5-52.5%, Spectrum™ Chemical
SDP

Catalog Number 18600878
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
C9H13ClN
170.66
[Cl-].C[N+](C)(*)CC1=CC=CC=C1
C9H13ClN
170.66
[Cl-].C[N+](C)(*)CC1=CC=CC=C1

B1068, 8001-54-5

Spectrum™ Chemical Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilizing agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 50%
UN Number UN2920
Quantity 200 L
Percent Purity 47.5 to 52.5%
Grade NF
Packaging Drum
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit