Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzamidine, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
618-39-3
C7H8N2
120.155
MFCD00042826
PXXJHWLDUBFPOL-UHFFFAOYSA-N
benzamidine,benzimidamide,phenylamidine,unii-kue3zy3j1f,ccris 2952,benzamidine protonated,kue3zy3j1f,chembl20936,1bra,1oss
2332
CHEBI:41033
benzenecarboximidamide
C1=CC=C(C=C1)C(=N)N
618-39-3
C7H8N2
120.155
MFCD00042826
PXXJHWLDUBFPOL-UHFFFAOYSA-N
benzamidine,benzimidamide,phenylamidine,unii-kue3zy3j1f,ccris 2952,benzamidine protonated,kue3zy3j1f,chembl20936,1bra,1oss
2332
CHEBI:41033
benzenecarboximidamide
C1=CC=C(C=C1)C(=N)N
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10g
Color Colorless to Yellow
Melting Point 65°C to 70°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 120.15
Physical Form Liquid
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material Benzamidine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.