Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzamil, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Na+/Ca2+ exhanger (NCX) inhibitor (IC50 ∽ 100 nM); TRPP3 channel blocker (IC50 = 1.1 μM for inhibition of Ca2+-activated TRPP3 channel activity). Also non-selective Deg/ENaC family blocker; reduces mechanosensitivity of colonic afferents. More potent derivative of amiloride (Cat. No. 0890).
Specifications
Quantity | 10 mg |
Formula Weight | 356.21 |
Percent Purity | >98% |
Chemical Name or Material | Benzamil |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction