Alfa Aesar™ Benzene-1,2-dithiocarboxamide, 97%

Manufacturer: Alfa Aesar™ H5183703

Quantity 1g

 View more versions of this product

Catalog No. AAH5183703

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Molecular Formula C8H8N2S2
    Formula Weight 196.28
    MDL Number MFCD16293897
    UN Number UN2811
    Chemical Name or Material Benzene-1,2-dithiocarboxamide

    Specifications

    Formula Weight 196.28
    Molecular Formula C8H8N2S2
    MDL Number MFCD16293897
    Quantity 1g
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P280e-P309-P310
    Wear protective clothing and face protection.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Benzene-1,2-dithiocarboxamide
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P280e-P309-P310
    Wear protective clothing and face protection.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit