Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Benzene-1,3-disulfonic acid disodium salt, 94%
Click to view available options
Quantity:
25g
100g
500g
Description
Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.
Solubility
Soluble in water( 663 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.
Solubility
Soluble in water( 663 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.124 |
Quantity | 25g |
Solubility Information | Soluble in water( 663g/L at 20°C). |
Formula Weight | 282.2 |
Percent Purity | 94% |
Chemical Name or Material | Benzene-1,3-disulfonic acid disodium salt |
Safety and Handling
EINECSNumber : 212-606-4
TSCA : Yes
Recommended Storage : Ambient temperatures