Alfa Aesar™ Benzene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 43817M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA43817M1

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 71-43-2
    Formula Weight 78.11g/mol
    MDL Number MFCD00003009
    UN Number UN1114
    Beilstein 969212
    Merck Index 14,1066
    Chemical Name or Material Benzene, Anhydrous

    Specifications

    Density 0.874g/mL
    Odor Aromatic
    Packaging ChemSeal™ bottles
    Refractive Index 1.5010
    Boiling Point 80°C
    Melting Point 5°C
    Quantity 4 x 1L
    CAS 71-43-2
    Physical Form Liquid
    Formula Weight 78.11g/mol
    MDL Number MFCD00003009
    UN Number UN1114
    Beilstein 969212
    Merck Index 14,1066
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H340-H350-H372-H304-H315-H319
    Highly flammable liquid and vapor.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: BENZENE
    RTECS Number CY1400000
    TSCA TSCA
    Chemical Name or Material Benzene, Anhydrous
    Assay 99.8%

    Description

    • Packaged under Argon

    GHS H Statement
    H225-H340-H350-H372-H304-H315-H319
    Highly flammable liquid and vapor.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit