Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzene-d6, 100% (Isotopic)

Manufacturer:  Alfa Aesar™ 42265SA

Quantity 2 each

 View more versions of this product

Catalog No. AA42265SA


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1076-43-3
MFCD00003010
Benzene-d_6, Isotopic
84.16
UN1114

Specifications

1076-43-3
Prescored Ampules
0.950g/mL
6.8°C
2 each
84.16
UN1114
100%
Aromatic
Moisture Sensitive
79.1°C
Liquid
1.4986
MFCD00003010
Benzene-d_6, Isotopic

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H340-H350-H372-H304-H315-H319
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: BENZENE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit