Alfa Aesar™ Benzene-d6, 99.5% (Isotopic)

Manufacturer: Alfa Aesar™ 8953009

Quantity 10g

 View more versions of this product

Catalog No. AA8953009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1076-43-3
    Formula Weight 84.16
    MDL Number MFCD00003010
    UN Number UN1114
    Chemical Name or Material Benzene-d_6, Isotopic

    Specifications

    Formula Weight 84.16
    Health Hazard 2 GHS H Statement
    H225-H340-H350-H372-H304-H315-H319
    Highly flammable liquid and vapor.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Boiling Point 79.1°C
    Physical Form Liquid
    MDL Number MFCD00003010
    UN Number UN1114
    Odor Aromatic
    TSCA TSCA
    Sensitivity Moisture Sensitive
    Density 0.950g/mL
    Quantity 10g
    Refractive Index 1.4986
    CAS 1076-43-3
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: BENZENE
    Melting Point 6.8°C
    Chemical Name or Material Benzene-d_6, Isotopic
    Assay 99.5%

    GHS H Statement
    H225-H340-H350-H372-H304-H315-H319
    Highly flammable liquid and vapor.
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit