Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzeneruthenium(II) chloride, dimer, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
37366-09-9
C12H12Cl4Ru2
500.18
MFCD00064686
YGXMUPKIEHNBNQ-UHFFFAOYSA-J
benzeneruthenium ii chloride dimer,dibenzenetetrachlorodiruthenium,bis benzene dichlororuthenium,benzene dichlororuthenium dimer,benzeneruthenium dichloride dimer,bis benzene tetrachlorodiruthenium,rucl2 benzene 2,dichloro benzene rutheniumdimer,benzene ruthenium cento chloride
10962144
benzene;dichlororuthenium
C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
37366-09-9
C12H12Cl4Ru2
500.18
MFCD00064686
YGXMUPKIEHNBNQ-UHFFFAOYSA-J
benzeneruthenium ii chloride dimer,dibenzenetetrachlorodiruthenium,bis benzene dichlororuthenium,benzene dichlororuthenium dimer,benzeneruthenium dichloride dimer,bis benzene tetrachlorodiruthenium,rucl2 benzene 2,dichloro benzene rutheniumdimer,benzene ruthenium cento chloride
10962144
benzene;dichlororuthenium
C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
Packaging Glass bottle
Melting Point 249.0°C
Infrared Spectrum Authentic
Color Gray
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Fieser 07,109; 09,167; 11,182; 14,136; 15,135; 16,130
Solubility Information Solubility in water: insoluble
Formula Weight 500.18
Physical Form Turnings
Percent Purity 97%
Chemical Name or Material Benzeneruthenium(II) chloride, dimer
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning