Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzeneseleninic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6996-92-5
C6H6O2Se
189.083
MFCD00002100
WIHKGDVGLJJAMC-UHFFFAOYSA-N
phenyl-selensaeure,benzene selenoic acid,seleninobenzoic acid,acmc-1bhr1,benzeneselenine acid,benzeneseleninic acid,chembl39469,benzeneseleninic acid, purum t
23427
benzeneseleninic acid
C1=CC=C(C=C1)[Se](=O)O
6996-92-5
C6H6O2Se
189.083
MFCD00002100
WIHKGDVGLJJAMC-UHFFFAOYSA-N
phenyl-selensaeure,benzene selenoic acid,seleninobenzoic acid,acmc-1bhr1,benzeneselenine acid,benzeneseleninic acid,chembl39469,benzeneseleninic acid, purum t
23427
benzeneseleninic acid
C1=CC=C(C=C1)[Se](=O)O
Quantity 5g
Packaging Glass bottle
Water 6% max. (K.F.)
Melting Point 120°C to 124°C
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5Se(O)OH
Fieser 08,28; 09,32; 10,22; 14,176; 15,167; 16,177; 06,240; 07,139; 08,29; 09,32; 10,22; 11,37; 15,18
Formula Weight 189.07
Physical Form Crystalline Powder or Needles
Percent Purity ≥98.5% (on dry substance)
Chemical Name or Material Benzeneseleninic acid, 99%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger