Alfa Aesar™ Benzenesulfinic acid sodium salt, 98%

Manufacturer: Alfa Aesar™ A1211836

Quantity 500g

 View more versions of this product

Catalog No. AAA1211836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 873-55-2
    Molecular Formula C6H5NaO2S
    Formula Weight 164.16
    MDL Number MFCD00013135
    Beilstein 3598405
    Synonym Sodium benzenesulfinate
    Chemical Name or Material Benzenesulfinic acid sodium salt

    Specifications

    Formula Weight 164.16
    Melting Point >300°C
    CAS 873-55-2
    TSCA TSCA
    Molecular Formula C6H5NaO2S
    Quantity 500g
    Synonym Sodium benzenesulfinate
    RTECS Number DA9135000
    MDL Number MFCD00013135
    Beilstein 3598405
    Chemical Name or Material Benzenesulfinic acid sodium salt
    Assay 98%

    Description

    Applications
    It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit