Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzenesulfinic acid sodium salt dihydrate, 97%

Manufacturer:  Alfa Aesar™ B2557622

Quantity 100g

 View more versions of this product

Catalog No. AAB2557622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

25932-11-0
200.19 (164.16 Anhydrous)
Sodium benzenesulfinate dihydrate
C6H5NaO2S·2H2O
MFCD00149638
Benzenesulfinic acid sodium salt dihydrate

Specifications

25932-11-0
>310°C
MFCD00149638
Sodium benzenesulfinate dihydrate
97%
C6H5NaO2S·2H2O
200.19 (164.16 Anhydrous)
100g
Benzenesulfinic acid sodium salt dihydrate

Safety and Handling

SDS & Safety Documents

RTECSNumber : DA9135000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit