Alfa Aesar™ Benzenesulfinic acid sodium salt dihydrate, 97%

Manufacturer: Alfa Aesar™ B2557636

Quantity 500g

 View more versions of this product

Catalog No. AAB2557636

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 25932-11-0
    Molecular Formula C6H5NaO2S·2H2O
    Formula Weight 200.19 (164.16 Anhydrous)
    MDL Number MFCD00149638
    Synonym Sodium benzenesulfinate dihydrate
    Chemical Name or Material Benzenesulfinic acid sodium salt dihydrate

    Specifications

    Formula Weight 200.19 (164.16 Anhydrous)
    Melting Point >310°C
    TSCA TSCA
    Molecular Formula C6H5NaO2S·2H2O
    MDL Number MFCD00149638
    Quantity 500g
    Synonym Sodium benzenesulfinate dihydrate
    RTECS Number DA9135000
    CAS 25932-11-0
    Chemical Name or Material Benzenesulfinic acid sodium salt dihydrate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit