Alfa Aesar™ Benzenesulfonamide, 98+%

Manufacturer: Alfa Aesar™ A108910I

Quantity 5000g

 View more versions of this product

Catalog No. AAA108910I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-10-2
    Molecular Formula C6H7NO2S
    Formula Weight 157.19
    MDL Number MFCD00007930
    Beilstein 1100566
    Chemical Name or Material Benzenesulfonamide

    Specifications

    Formula Weight 157.19
    Molecular Formula C6H7NO2S
    Health Hazard 1 Warning
    Melting Point 151° to 154°C
    Flash Point 250°C (482°F)
    RTECS Number DA9380000
    TSCA TSCA
    MDL Number MFCD00007930
    CAS 98-10-2
    Beilstein 1100566
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Quantity 5000g
    Chemical Name or Material Benzenesulfonamide
    Assay ≥98%

    Description

    Applications
    The presence of the benzenesulfonylamido (BS) groups shifted the activity of classic antimicrobial fluoroquinolones from being more active against Gram-negative to Gram-positive strains. Biospecific adsorption of carbonic anhydrase to self-assembled monolayers of alkanethiolates that present benzenesulfonamide groups on gold. Biospecific binding of carbonic anhydrase to mixed sams presenting benzenesulfonamide ligands led to a model system for studying lateral steric effects. In engineering the specificity of antibacterial fluoroquinolones, benzenesulfonamide modifications at c-7 of ciprofloxacin change its primary target instreptococcus pneumoniae from topoisomerase iv to gyrase. Polar substitutions in the benzenesulfonamide ring of celecoxib afford a potent 1,5-diarylpyrazole class of COX-2 inhibitors.

    Solubility
    Soluble in methanol (25 mg/ml), and water (4.3 mg/ml at 16°C).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit