Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonamide +98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
98-10-2
C6H7NO2S
157.19
MFCD00007930
KHBQMWCZKVMBLN-UHFFFAOYSA-N
benzenesulphonamide,benzosulfonamide,benzolsulfonamide,phenyl sulfonamide,benzene sulfonamide,phenylsulfonamide,m and b 7973,chembl27601,benzolsulfonamid,benzensulfonamide
7370
benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)N
98-10-2
C6H7NO2S
157.19
MFCD00007930
KHBQMWCZKVMBLN-UHFFFAOYSA-N
benzenesulphonamide,benzosulfonamide,benzolsulfonamide,phenyl sulfonamide,benzene sulfonamide,phenylsulfonamide,m and b 7973,chembl27601,benzolsulfonamid,benzensulfonamide
7370
benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)N
Flash Point 250°C
Melting Point 151°C to 153°C
Infrared Spectrum Authentic
Quantity 100g
Packaging Plastic bottle
Color White
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H5SO2NH2
Beilstein 11, 39
Solubility Information Solubility in water: 4.3g/L (16°C)
Formula Weight 157.19
Physical Form Powder, Crystals and/or Chunks
Percent Purity ≥98.0%
Chemical Name or Material Benzenesulfonamide, 98+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-637-1

RTECSNumber : DA9380000

TSCA : TSCA